Learn More
Bochem™ Aluminum Rod for Stand Base M10
The aluminum rods with M10 thread for used with Bochem stand bases and clamps. For holding and supporting laboratory apparatus.
Brand:  Bochem™ 5133
Product Code. 10320631
  • 18.45 € / Each
Estimated Shipment: 18-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Aluminum
  • Thread M10
Specifications
Specifications
Aluminum
12 mm
Holdaing and Supporting Laboratory Apparatus
1000 mm
Aluminum Stand Rod M10
Stand Base M10, Clamps