missing translation for 'onlineSavingsMsg'
Learn More
Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals Product Code.: 10744124

Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals

Product Code. 10744124
2.5 kg, Plastic bottle
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Glass Bottle
Plastic bottle

Product Code. 10744124

Brand: Thermo Scientific Acros 400590025

Please to purchase this item. Need a web account? Register with us today!

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-31-8
Molecular Formula Al2O12S3·18H2O
Molecular Weight (g/mol) 666.4
MDL Number MFCD00149136
InChI Key AMVQGJHFDJVOOB-UHFFFAOYSA-H
Synonym aluminum sulfate octadecahydrate, patent alum, cake alum octadecahydrate, unii-tcs9l00g8f, dialuminum sulfate octadecahydrate, tcs9l00g8f, aluminum sulfate.18h2o, aluminum sulfate.18h2o, dialuminium sulfate octadecahydrate, aluminum sulfate-water 1/18Show MoreShow Less
PubChem CID 22377415
ChEBI CHEBI:74779
IUPAC Name dialuminum;trisulfate;octadecahydrate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[Al+3]

Specifications

Melting Point 86.0°C
Color White
Physical Form Crystalline Powder
Quantity 2.5 kg
Assay Percent Range 98.0 to 102% (Complexometry)
Linear Formula Al2(SO4)3·18H2O
Merck Index 15, 360
Solubility Information Solubility in water: soluble. Other solubilities: practically insoluble in alcohol
Formula Weight 666.4
Percent Purity ≥98%
Grade ACS Reagent
Packaging Plastic bottle
Chemical Name or Material Aluminum sulfate octadecahydrate
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0460132 Certificate TypeCertificate of Analysis Date09/02/2024 Catalog Number400590025
Lot NumberA0399032 Certificate TypeCertificate of Analysis Date14/12/2022 Catalog Number
Lot NumberA0390340 Certificate TypeCertificate of Analysis Date13/12/2022 Catalog Number
Lot NumberA0442456 Certificate TypeCertificate of Analysis Date23/05/2022 Catalog Number
Lot NumberA0431654 Certificate TypeCertificate of Analysis Date23/06/2021 Catalog Number
compliance-icons
Product Identifier
  • Aluminium sulfate octadecahydrate
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H318-Causes serious eye damage.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P234-Keep only in original container.
  • P390-Absorb spillage to prevent material damage.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum sulfate octadecahydrate, 98+%, ACS reagent, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.