missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Aluminum Scaffolding Tubes
Provides for sturdy scaffolding construction in the laboratory. Fisherbrand™ Aluminum Scaffolding Tubes are manufactured from a durable aluminum alloy.
9.60 € - 17.30 €
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
11562662
|
Fisherbrand™
2121/1 |
1 m |
9.60 €
Each |
Estimated Shipment: 13-03-2025
Log in to see stock. |
|||||
11572662
|
Fisherbrand™
2121/2 |
2 m |
17.30 €
Each |
Estimated Shipment: 10-01-2025
Log in to see stock. |
|||||
Description
- Approx. 13mm diameter
- Available in two different lengths
- Fits with Fisherbrand scaffolding connectors and foot adapters
Specifications
Aluminum Alloy | |
Tube |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title