missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Aluminum Anodized Test Tube Racks

Wesbart aluminum anodized racks to store test tubes

Brand:  Wesbart™ SP2384

Code : A0

Additional Details : Weight : 0.80000kg

 View more versions of this product

Product Code. 11751724

  • 59.55 € / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Single, double, or triple platform
  • aluminum test tube racks to store tubes.
Specifications

Specifications

Tube Rack
Aluminum
341 x 200 x 30mm
Tubes (23.5 mm)
84
23.5 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers