missing translation for 'onlineSavingsMsg'
Learn More

Wesbart™ Aluminum Anodized Test Tube Racks

Wesbart aluminum anodized racks to store test tubes

Brand:  Wesbart™ SP2036

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 17276904

  • 34.85 € / Each
Estimated Shipment: 09-08-2024
to see stock.
Add to basket

Description

Description

  • Single, double, or triple platform
  • aluminum test tube racks to store tubes.
Specifications

Specifications

Test Tube Rack
Autoclavable
Tubes (20 mm)
Rectangular
Non-sterile
30 mm
Aluminum Test Tube Rack
Silver
36
Yes
165 x 165 x 30 mm
20 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers