Learn More
Aluminum bromide, 98%, Thermo Scientific Chemicals
32.30 € - 146.73 €
Chemical Identifiers
CAS | 7727-15-3 |
---|---|
Molecular Formula | AlBr3 |
Molecular Weight (g/mol) | 266.69 |
MDL Number | MFCD00003421 |
InChI Key | PQLAYKMGZDUDLQ-UHFFFAOYSA-K |
Synonym | Aluminum tribromide |
IUPAC Name | aluminium(3+) tribromide |
SMILES | [Al+3].[Br-].[Br-].[Br-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316138
|
Thermo Scientific Alfa Aesar
011113.09 |
10 g | N/A | ||||||
11306138
|
Thermo Scientific Alfa Aesar
011113.22 |
100 g |
146.73 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alkylation, bromination and isomerization catalyst in organic synthesis.Aluminum bromide acts as a strong Lewis acid and is used in Lewis acid-promoted reactions such as epoxide ring openings. It is used as a catalyst for Friedel-Crafts alkylation reaction. It also finds application in bromination and isomerization reactions in organic synthesis. It is also used in polymerization reaction to prepare poly(o-xylene)from benzocyclobutene.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7727-15-3 | |
266.69 | |
PQLAYKMGZDUDLQ-UHFFFAOYSA-K | |
aluminium(3+) tribromide |
AlBr3 | |
MFCD00003421 | |
Aluminum tribromide | |
[Al+3].[Br-].[Br-].[Br-] |
Specifications
265°C | |
7727-15-3 | |
Crystalline | |
AlBr3 | |
Aluminum tribromide | |
PQLAYKMGZDUDLQ-UHFFFAOYSA-K | |
aluminium(3+) tribromide | |
266.71 | |
Moisture sensitive | |
3.21 g/cm3 |
97.5°C | |
Orange to White | |
98% | |
MFCD00003421 | |
Reacts with water violently,soluble in alcohol,carbon disulfide,ether.Slightly soluble in methanol,diethyl ether and acetone. | |
[Al+3].[Br-].[Br-].[Br-] | |
266.69 | |
Pungent | |
Reacts with water violently | |
Aluminum bromide |
Safety and Handling
Keep container dry. When using do not eat or drink. In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Never add water to this product. Wear suitable protective clothing, gloves and eye/face protection. In case of accident or if you feel unwell, seek medical advice immediately (show the label where possible). This material and/or its container must be disposed of as hazardous waste.
P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
H302-H314
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 231-779-7
missing translation for 'rtecsNumber' : BD0350000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
- Aluminum bromide
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.