missing translation for 'onlineSavingsMsg'
Learn More

Aluminum bromide, 99.997% (metals basis)

Product Code. p-4781758
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11339379

Brand: Thermo Scientific Alfa Aesar 089065.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7727-15-3 | AlBr3 | 266.69 g/mol

Aluminum Bromide, is used as a catalyst for the Friedel-Crafts alkylation reaction, and also for bromination and isomerization in organic synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7727-15-3
Molecular Formula AlBr3
Molecular Weight (g/mol) 266.69
MDL Number MFCD00003421
InChI Key PQLAYKMGZDUDLQ-UHFFFAOYSA-K
Synonym Aluminum tribromide
IUPAC Name aluminium(3+) tribromide
SMILES [Al+3].[Br-].[Br-].[Br-]

Specifications

Boiling Point 265°C
Melting Point 97.5°C
Color Orange to White
Physical Form Crystalline
Quantity 5 g
Assay Percent Range 99.997% (Metals basis)
Solubility Information Soluble in benzene,alcohol,carbon disulfide,ether and toluene. Reacts with water.
Formula Weight 266.71
Odor Pungent
Sensitivity Moisture sensitive
Density 3.21 g/cm3
Chemical Name or Material Aluminum bromide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminum bromide
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • EUH014-Reacts violently with water.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Aluminum bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.