missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum calcium isopropoxide, 99.8% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals
Click to view available options
Quantity:
25 mL
100 mL
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Spécification
Boiling Point | 230°C to 240°C (4.0 mmHg) |
Odor | Alcohol-like |
Quantity | 25 mL |
UN Number | UN1219 |
Sensitivity | Moisture sensitive |
Formula Weight | 566.3 |
Percent Purity | 99.8% |
Physical Form | Liquid |
Assay | (metals basis) |
Chemical Name or Material | Aluminum calcium isopropoxide |
Sécurité et traitement
Identifiant produit
- Aluminum calcium isopropoxide 10% w/v in isopropanol
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'