missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Aluminum RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand: Sperian™ 1783040
This item is not returnable.
View return policy
Description
- Body made of aluminum provides optimum resistance to hazards
- Low breathing resistance
- Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
Specifications
Canister | |
RD40 | |
ALU RD40 A2B2P3 | |
Full-Face mask equipped with an Rd40 connector. | |
1 Ea. |
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7752 A/13/1 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7752 A/13/1 PSA, Certifications- EN 14387, EN 143, EN 12941, HON filter | |
Aluminum alloy with PE cover activated and impregnated charcoal. | |
Respiratory protection against organic gases and vapors with a boiling point below 65°C, inorganic gases and vapors, solid and liquid particles | |
Model A2B2P3RD |