Learn More
Sperian™ Honeywell™ Aluminum RD40 Filters
Protects against a multitude of risks encountered in many environments. Sperian™ Honeywell™ Aluminum RD40 Filters are positioned low to prevent obstruction in the field of vision.
Brand:  Sperian™ 1784000
 View more versions of this product
Product Code. 11897040
  • 73.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Body made of aluminum provides optimum resistance to hazards
  • Low breathing resistance
  • Available in models to protect against airborne particulates, biohazards, chemicals, contamination and gas, vapors and smoke
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Canister
RD40
ALU RD40 A2B2E2K2P3
Full-face mask equipped with an Rd40 connector
1 Ea.
E.C. Declaration of Conformity- EC Category PPE- 3, Quality Assurance- ISO 9001 / 2000, EC Certificate Number- 7549 A/12/23 PSA, EC Attestation- EC Attestation, EC Attestation Number- 7549 A/12/23 PSA, Certifications- EN 14387, EN 148, EN 12941, HON filte
Aluminum alloy with PE cover activated and impregnated charcoal
Respiratory protection against ammonia and ammonia derivatives, solid and liquid particles
Model ABEK2P3RD