missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Haldenwanger™ Aluminum Ceramic Cylindrical Flat Bottom Crucibles
24.80 € - 121.00 €
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
10540252
|
Haldenwanger™
L ALSINTTIEGEL*A/1 |
5 mL |
24.80 €
Each |
Estimated Shipment: 29-10-2024
Log in to see stock. |
|||||
10252311
|
Haldenwanger™
L ALSINTTIEGEL*A/3 |
30 mL |
53.75 €
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10453731
|
Haldenwanger™
L ALSINTTIEGEL*A/4 |
60 mL |
71.95 €
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
10048660
|
Haldenwanger™
L ALSINTTIEGEL*A/5 |
110 mL |
121.00 €
Each |
Estimated Shipment: 15-10-2024
Log in to see stock. |
|||||
Specifications
Aluminum Ceramic | |
Crucible | |
Cylinder |