missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, anhydrous, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11400800
Click to view available options
Quantity:
5 g
25 g
100 g

It is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Melting Point 190°C (sublimation)
pH 2.4
Quantity 25 g
UN Number UN1726
Merck Index 14,337
Solubility Information Soluble in benzene,carbon tetrachloride,chloroform,hydrochloric acid,ether,benzophenone,nitrobenzene and ethanol.
Formula Weight 133.34
Percent Purity 99.999%
Odor Pungent
Sensitivity Moisture sensitive
Density 2.44 g/mL
Assay (metals basis)
Chemical Name or Material Aluminum chloride, Anhydrous
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'