missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride hexahydrate, Puratronic™, 99.9995% (metals basis), Thermo Scientific Chemicals

25.33 € - 580.89 €

Chemical Identifiers

CAS 7784-13-6
Molecular Formula AlCl3H12O6
Molecular Weight (g/mol) 241.42
MDL Number MFCD00149134
InChI Key JGDITNMASUZKPW-UHFFFAOYSA-K
IUPAC Name aluminium(3+) hexahydrate trichloride
SMILES O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11350799
View Documents
Thermo Scientific Alfa Aesar
010622.06
5 g
25.33 €
5g
Please to purchase this item. Need a web account? Register with us today!
11370799
View Documents
Thermo Scientific Alfa Aesar
010622.18
50 g
135.68 €
50g
Please to purchase this item. Need a web account? Register with us today!
11380799
View Documents
Thermo Scientific Alfa Aesar
010622.30
250 g
580.89 €
250g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Pharmaceutical preparations of aluminum chloride hexahydrate are used to treat hyperhidrosis, a condition involving excessive sweating. It is also employed in deodorants, antiperspirant preparations, dyeing fabrics, refining crude oil, textile finishing, manufacturing parchment paper, preserving the wood and disinfecting stables. It also finds use as a reagent in organic synthesis. Aluminum chloride hexahydrate is also useful in water treatment process, medicine and cosmetic products.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7784-13-6
241.42
JGDITNMASUZKPW-UHFFFAOYSA-K
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
AlCl3H12O6
MFCD00149134
aluminium(3+) hexahydrate trichloride

Specifications

100°C (decomposition)
White
Crystalline Aggregates
MFCD00149134
14,337
JGDITNMASUZKPW-UHFFFAOYSA-K
aluminium(3+) hexahydrate trichloride
241.43 (133.34 Anhydrous)
Puratronic™
2.398 g/mL
Aluminum chloride hexahydrate
7784-13-6
2.5
AlCl3H12O6
UN3260
Highly soluble in water. Soluble in alcohol,ether,glycerol,propylene glycol and acetonitrile.
O.O.O.O.O.O.[Al+3].[Cl-].[Cl-].[Cl-]
241.42
99.9995%
Odorless
(metals basis)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 231-208-1

missing translation for 'rtecsNumber' : BD0530000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Aluminum chloride hexahydrate

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 C

Hazard Statement
  • H314-Causes severe skin burns and eye damage.

Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.