missing translation for 'onlineSavingsMsg'
Learn More

Aluminum chloride, Anhydrous, sublimed, ≥98%, Honeywell Fluka™

Product Code. 15651000
Click to view available options
Quantity:
500 g
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Anhydrous, sublimed,≥98%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-70-0
Molecular Formula AlCl3
Molecular Weight (g/mol) 133.33
MDL Number MFCD00003422
InChI Key VSCWAEJMTAWNJL-UHFFFAOYSA-K
Synonym aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum
IUPAC Name aluminum(3+) trichloride
SMILES [Al+3].[Cl-].[Cl-].[Cl-]

Specifications

Physical Form Anhydrous, Sublimed
Quantity 500 g
Linear Formula AlCl3
UN Number 1726
Formula Weight 133.34g/mol
Percent Purity ≥98%
Vapor Pressure 1mmHg (100°C)
Chemical Name or Material Aluminum Chloride
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'