Learn More
Aluminum chloride, ultra dry, 99.99% (metals basis), Thermo Scientific Chemicals
84.70 € - 593.00 €
Chemical Identifiers
CAS | 7446-70-0 |
---|---|
Molecular Formula | AlCl3 |
Molecular Weight (g/mol) | 133.33 |
MDL Number | MFCD00003422 |
InChI Key | VSCWAEJMTAWNJL-UHFFFAOYSA-K |
Synonym | aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum |
IUPAC Name | aluminum(3+) trichloride |
SMILES | [Al+3].[Cl-].[Cl-].[Cl-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11362498
|
Thermo Scientific Alfa Aesar
014552.06 |
5 g |
84.70 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11342498
|
Thermo Scientific Alfa Aesar
014552.14 |
25 g |
215.00 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11352498
|
Thermo Scientific Alfa Aesar
014552.22 |
100 g |
593.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum chloride is used to control excessive sweating. It is commonly used as a catalyst for the halogenation (especially chlorination) of aromatic groups, as well as in the Friedel Crafts reaction. It is also used in the Meerwein-Ponndorf-Verley reduction. It is widely used in the manufacturing of petrochemicals such as alkylbenzene, ethylbenzene, alkyl aryl ketone, ethyl chloride. It is used for making detergents, polymerization and isomerization reactions of hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7446-70-0 | |
133.33 | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride |
AlCl3 | |
MFCD00003422 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
[Al+3].[Cl-].[Cl-].[Cl-] |
Specifications
190° (sublimation) | |
Gray to Yellow | |
99.99% (Metal basis) | |
MFCD00003422 | |
Soluble in benzene,carbon tetrachloride,chloroform,hydrochloric acid,ether,benzophenone,nitrobenzene and ethanol. | |
[Al+3].[Cl-].[Cl-].[Cl-] | |
133.33 | |
Pungent | |
Yellow to gray powder | |
Aluminum chloride, ultra dry |
7446-70-0 | |
Powder | |
AlCl3 | |
aluminum chloride anhydrous, aluminum iii chloride, aluminiumchloride, aluminumchloride, aluminum chlorid, aluminum cloride, alumimum chloride, aluminum-chloride, trichloroaluminium, chloride aluminum | |
VSCWAEJMTAWNJL-UHFFFAOYSA-K | |
aluminum(3+) trichloride | |
133.34 | |
Moisture sensitive | |
2.44 g/cm3 |
Safety and Handling
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : DOT Class: 8, Packaging Group: II
missing translation for 'einecsNumber' : 231-208-1
missing translation for 'rtecsNumber' : BD0525000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum chloride, ultra dry
Signal Word
- Danger
Hazard Category
- Skin corrosion/irritation Category 1 B
Hazard Statement
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
- P363-Wash contaminated clothing before reuse.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only