missing translation for 'onlineSavingsMsg'
Learn More

Bochem™ Aluminum Epoxy Lab jack

Manufactured according to DIN 12897. Bochem™ Aluminum Epoxy Lab jack features improved scissor technology with reduced tolerances.

Brand:  Bochem™ 11030

 View more versions of this product

Product Code. 10439870

  • 185.00 € / Each
Estimated Shipment: 14-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Improved scissor technology with reduced tolerances
  • Ergonomic knob, rounded and user friendly
  • Scissor-construction made of 18/10-stainless steel
  • Manufactured according to DIN 12897
  • Maximal dynamic operating load (Kg max. dyn.) and maximal static operating load (Kg max. stat.) norms according to DIN 12897
  • Smaller models (up to 240x240mm; Typ 1) have a simple hand-operated adjusting wheel
  • Bigger models (from 300x300mm; Typ 2) use of included ratchet, which connects with a special adjusting wheel, is recommended
Specifications

Specifications

Aluminum Epoxy
200 mm
1 Ea.
60 to 275 mm
Lab Jack
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bochem™ Aluminum Epoxy Lab jack > Length: 200mm

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.