Learn More
Bochem™ Aluminum Epoxy Lab jack
Manufactured according to DIN 12897. Bochem™ Aluminum Epoxy Lab jack features improved scissor technology with reduced tolerances.
Brand:  Bochem™ 11040
 View more versions of this product
Product Code. 10645991
  • 208.00 € / Each
Estimated Shipment: 25-02-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Improved scissor technology with reduced tolerances
  • Ergonomic knob, rounded and user friendly
  • Scissor-construction made of 18/10-stainless steel
  • Manufactured according to DIN 12897
  • Maximal dynamic operating load (Kg max. dyn.) and maximal static operating load (Kg max. stat.) norms according to DIN 12897
  • Smaller models (up to 240x240mm; Typ 1) have a simple hand-operated adjusting wheel
  • Bigger models (from 300x300mm; Typ 2) use of included ratchet, which connects with a special adjusting wheel, is recommended
Specifications
Specifications
Aluminum Epoxy
240 mm
1 Ea.
60 to 275 mm
Lab Jack