missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eppendorf™ Aluminum Fixed Angle Waterproof Rotor
Choice of four different rotors
Brand: Eppendorf™ 5424700004
This item is not returnable.
View return policy
Description
- Choice of four different rotors
- Standard rotor can accommodate 1.5mL/2.0mL tubes
- PCR* strips and kits/spin columns can be centrifuged in the corresponding rotor For molecular and biological laboratories.
Specifications
Aluminum | |
24 x 1.5/2 mL | |
Aerosol rotor lid |
Fixed Angle Rotor | |
14,680 rpm |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction