missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum flake, APS 11 micron, 99.7% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
250 g
1 kg
5 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Color | Silver to Gray |
Appearance | Silver Gray flakes |
Density | 2.7g/cm3 |
Assay Percent Range | 99.7% (Metals basis) |
Quantity | 1 kg |
Ignition Point | 400°C (752°F) |
IUPAC Name | aluminium |
Odor | Odorless |
Sensitivity | Water reactive |
Form | Flake |
Show More |
Safety and Handling
Product Identifier
- Aluminum flake
- Danger
- Flammable solid Category 1
- Substance which\, in contact with water\, emits flammable gases Category 2
- H228-Flammable solid.
- H261-In contact with water releases flammable gases.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P231+P232-Handle under inert gas. Protect from moisture.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P370+P378-In case of fire: Use dry sand, dry chemical or alcohol-resistant foam to extinguish.
- MIXTURE LIST-Contains : Aluminum
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'