missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride hydrate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11390799
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 32287-65-3
Molecular Formula AlF3H2O
Molecular Weight (g/mol) 101.99
MDL Number MFCD00149142
InChI Key XTXFDUABTPNTFB-UHFFFAOYSA-K
Synonym aluminum fluoride hydrate, alf3.h2o, trifluoroalumane-water 1/1, aluminum fluoride hydrate, puratronic
PubChem CID 16211462
IUPAC Name trifluoroalumane hydrate
SMILES O.F[Al](F)F

Specifications

Physical Form Powder
Quantity 5 g
UN Number UN3260
Formula Weight 83.98 (Anhydrous)
Percent Purity 99.99%
Grade Puratronic™
Assay (metals basis)
Chemical Name or Material Aluminum fluoride hydrate
compliance-icons compliance-icons
Product Identifier
  • Aluminum fluoride hydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H301-Toxic if swallowed.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'