missing translation for 'onlineSavingsMsg'
Learn More

Aluminum fluoride trihydrate, 97%, Thermo Scientific Chemicals

Product Code. 11367167
Click to view available options
Quantity:
100 g
500 g
2 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum fluoride trihydrate is used as an additive for the production of aluminum by electrolysis. It is also used to inhibit fermentation and in the preparation of low index films. It serves as a flux ingredient for the removal of magnesium in the refining of aluminum scrap. Further, it is used in the manufacture of aluminum silicates, ceramics and glass. In addition, it acts as a catalyst for organic synthesis and filler in glass industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15098-87-0
Molecular Formula AlF3H6O3
Molecular Weight (g/mol) 138.022
MDL Number MFCD00003426
InChI Key BKLZIAYVINRQEJ-UHFFFAOYSA-K
Synonym aluminum trifluoride trihydrate, aluminum 3+ trihydrate trifluoride, aluminum fluoride-water 1/3/3, aluminum fluoride, trihydrate,, aluminum 3+ ion trihydrate trifluoride, fluoride, fluoride, aluminum salt, hydrate, hydrate, hydrate, fluoride
PubChem CID 22002992
IUPAC Name aluminum;trifluoride;trihydrate
SMILES O.O.O.[F-].[F-].[F-].[Al+3]

Specifications

Melting Point 250°C (in -3H2O)
Color White
Physical Form Powder
Quantity 2 kg
Merck Index 14,339
Solubility Information Slightly soluble in water,acids and alkalis. Insoluble in alcohol and acetone.
Formula Weight 138.02 (83.98 Anhydrous)
Percent Purity 97%
Odor Odorless
Density 2.17 g/mL
Chemical Name or Material Aluminum fluoride trihydrate
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'