missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Foil, 0.1mm (0.004 in.) thick, Puratronic™, 99.997% (Metals basis), Thermo Scientific Chemicals
Form: Approx. 2.7g/100 x 100mm; Temper: as rolled
109.55 € - 1014.05 €
Chemical Identifiers
CAS | 7429-90-5 |
---|---|
Molecular Formula | Al |
Molecular Weight (g/mol) | 26.98 |
MDL Number | MFCD00134029 |
InChI Key | XAGFODPZIPBFFR-UHFFFAOYSA-N |
Synonym | aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum |
PubChem CID | 5359268 |
ChEBI | CHEBI:33629 |
SMILES | [Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15435975
|
Thermo Scientific Alfa Aesar
011376.IP |
5 x 100 x 400 mm |
1014.05 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11391159
|
Thermo Scientific Alfa Aesar
011376.GH |
100 x 100 mm |
109.55 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11301169
|
Thermo Scientific Alfa Aesar
011376.GL |
100 x 400 mm |
283.41 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7429-90-5 | |
26.98 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
5359268 | |
[Al] |
Al | |
MFCD00134029 | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
CHEBI:33629 |
Specifications
Silver | |
Silver-colored foil | |
Al | |
aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum | |
[Al] | |
26.98 | |
CHEBI:33629 | |
Foil |
7429-90-5 | |
99.997% (Metals basis) | |
MFCD00134029 | |
XAGFODPZIPBFFR-UHFFFAOYSA-N | |
aluminum | |
5359268 | |
Odorless | |
Aluminum |
Safety and Handling
missing translation for 'einecsNumber' : 231-072-3
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only