missing translation for 'onlineSavingsMsg'
Learn More

Aluminum foil, 0.2mm (0.008in) thick, annealed, Puratronic, 99.9999% (metals basis), Thermo Scientific Chemicals

Product Code. 11458450
Click to view available options
Quantity:
100 x 500 mm
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Identificadores de productos químicos

CAS 7429-90-5
Molecular Formula Al
Molecular Weight (g/mol) 26.98
MDL Number MFCD00134029
InChI Key XAGFODPZIPBFFR-UHFFFAOYSA-N
Synonym aluminum, metal, powder, aluminio, metana, adom, alumina fibre, aluminum flake, dust, noral aluminum
PubChem CID 5359268
ChEBI CHEBI:33629
SMILES [Al]

Especificaciones

Quantity 100 x 500 mm
IUPAC Name aluminum
Percent Purity 99.9999%
Grade Puratronic™
Odor Odorless
Weight ≈5.4g/100x100mm
Form Foil
Assay (metals basis)
Chemical Name or Material Aluminum foil, 0.2mm (0.008 in.) thick, Annealed, Temper: soft

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'