Learn More
Welch™ Aluminum KF Inner Centering with Viton O-ring
Designed for use with CRVpro Rotary Vane Vacuum Pumps. Welch Ilmvac™ Aluminum KF Inner Centering Rings feature a replaceable Fluoroelastomer O-ring seal.
6.80 € - 11.13 €
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With | Price | Quantity & Availability | |||||
10680343
|
Welch™
701091 |
Separator 16 DN |
6.80 €
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
10152522
|
Welch™
701093 |
Separator 25 DN |
8.48 €
Each |
Estimated Shipment: 21-11-2024
Log in to see stock. |
|||||
10090992
|
Welch™
701095 |
Separator 40 DN |
11.13 €
Each |
Estimated Shipment: 08-11-2024
Log in to see stock. |
|||||
Description
- Stainless steel centering ring with Viton O-ring
- Replaceable Fluoroelastomer O-ring seal
- Centering ring assemblies are placed between two ISO flanges with matching outer diameters
- he widest portion of the centering ring rests inside a centering ring locater groove (blank flange) or counterbore (bored flange) on the Kwik-Flange™
- The O-ring rests on the flat polished surface outside the locater groove or counterbore
Specifications
Aluminum | |
1 Each | |
KF Inner Centering |