missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum Magnesium gauze, alloy 5056, 30 mesh woven from 0.23mm (0.009 in.) dia. wire, Thermo Scientific Chemicals
95.27 € - 469.00 €
Specifications
Chemical Name or Material | Aluminum Magnesium gauze, Alloy 5056 |
---|---|
Name Note | Woven from 0.23mm (0.009 in.) dia. wire |
Avg. Mol. Wt. or Mol. Wt. Range | Al:Mg; 95:5 wt% |
MDL Number | MFCD00214039 |
Mesh Size | 30 Mesh |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11305187
|
Thermo Scientific Alfa Aesar
040906.FL |
75 x 75 mm |
95.27 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11395177
|
Thermo Scientific Alfa Aesar
040906.GT |
150 x 150 mm |
186.93 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11315187
|
Thermo Scientific Alfa Aesar
040906.HB |
300 x 300 mm | N/A | ||||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Aluminum Magnesium gauze, Alloy 5056 | |
Al:Mg; 95:5 wt% | |
30 Mesh | |
Wire Cloth |
Woven from 0.23mm (0.009 in.) dia. wire | |
MFCD00214039 | |
Odorless |
Safety and Handling
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only