missing translation for 'onlineSavingsMsg'
Learn More

Aluminum magnesium isopropoxide, 99.9% (metals basis), 10% w/v in isopropanol, Thermo Scientific Chemicals

Product Code. 11388028
Click to view available options
Quantity:
25 mL
100 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

Aluminum magnesium isopropoxide has been used to study the hydrolysis conditions on the properties of magnesium aluminate spinel powders.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69207-83-6
MDL Number MFCD00070426
Synonym Magnesium aluminum isopropoxide

Specifications

Color Colorless
Boiling Point 130°C to 142°C (2.0 mmHg)
Quantity 25 mL
UN Number UN1219
Sensitivity Moisture sensitive
Solubility Information Not miscible with water.
Formula Weight 550.53
Percent Purity 99.9%
Physical Form Liquid
Assay (metals basis)
Chemical Name or Material Aluminum magnesium isopropoxide
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Aluminum magnesium isopropoxide 10% w/v in isopropanol
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.