Learn More
Sampling Systems™ Aluminum Multilevel Slot Sampler
Constructed of aluminum, making it suitable for sampling free flowing powders, granules and even slightly cohesive powders. Sampling Systems™ Aluminum Multilevel Slot Sampler is for taking a large volume, cross sectional sample from a container.
Brand:  Sampling Systems™ 1210J-2500
 View more versions of this product
Product Code. 15859221
  • 365.00 € / Each
Estimated Shipment: 30-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Made from aluminum
  • Cross section (Composite sample)
  • Suitable for taking large volume samples
  • aluminum samplers are not suitable for use with GMP products
Specifications
Specifications
Aluminum
7 Slots
50 mm
Slot Sampler
2.9 L
2500 mm