missing translation for 'onlineSavingsMsg'
Learn More

Aluminum n-butoxide, 90+%, Thermo Scientific Chemicals

Product Code. 11355029
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3085-30-1
Molecular Formula C12H30AlO3
Molecular Weight (g/mol) 249.351
MDL Number MFCD00015237
InChI Key YGVBJFIUURLJDC-UHFFFAOYSA-N
Synonym aluminum iii n-butoxide, aluminum; butan-1-ol
PubChem CID 50933522
IUPAC Name aluminum;butan-1-ol
SMILES CCCCO.CCCCO.CCCCO.[Al]

Specifications

Melting Point 102°C
Physical Form Lump
Quantity 10 g
Linear Formula Al[O(CH2)3CH3]3
UN Number UN3181
Formula Weight 246.32
Percent Purity ≥90%
Sensitivity Moisture sensitive
Chemical Name or Material Aluminum n-butoxide

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'