missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ 3-Prong Utility Clamp

With a screwed shaft and nickel-plated bosshead which can be screwed up and down shaft 35mm and locked in any position with a wing nut.

Brand:  Fisherbrand™ 1029

Product Code. 12926826

  • 141.80 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

  • With jaws covered in chemically resistant plastic for a safe and secure hold
  • Bosshead accepts rods up to 13mm
  • Rods are secured with a stainless steel thumb screw
Specifications

Specifications

1 Ea.
Utility Clamp
Aluminum Nickel, Plastic
90 mm Max.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ 3-Prong Utility Clamp > Material: Aluminum Nickel/Plastic

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.