Learn More
Aluminum nitrate hydrate, Puratronic™, 99.999% (metals basis excluding Hg), Thermo Scientific Chemicals
36.88 € - 700.49 €
Chemical Identifiers
CAS | 25838-59-9 |
---|---|
Molecular Formula | AlN3O9 |
Molecular Weight (g/mol) | 212.99 |
MDL Number | MFCD00003419 |
InChI Key | JLDSOYXADOWAKB-UHFFFAOYSA-N |
Synonym | aluminum nitrate hydrate, al.3no3.h2o, aluminum nitrate hydrate, puratronic, aluminum 3+ hydrate trinitrate |
PubChem CID | 22078336 |
SMILES | [Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11330809
|
Thermo Scientific Alfa Aesar
010626.06 |
5 g |
36.88 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11340809
|
Thermo Scientific Alfa Aesar
010626.14 |
25 g |
88.04 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11320809
|
Thermo Scientific Alfa Aesar
010626.22 |
100 g |
255.27 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11350809
|
Thermo Scientific Alfa Aesar
010626.36 |
500 g |
700.49 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum nitrate hydrate is used to produce alumina for the preparation of insulating papers, and in the extraction of actinide elements. It is also used in the preparation of rare earth aluminates, in the heating elements of cathode ray tubes, as a precursor for sol-gel synthesis of alumina, and on transformer core laminates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
25838-59-9 | |
212.99 | |
JLDSOYXADOWAKB-UHFFFAOYSA-N | |
22078336 |
AlN3O9 | |
MFCD00003419 | |
aluminum nitrate hydrate, al.3no3.h2o, aluminum nitrate hydrate, puratronic, aluminum 3+ hydrate trinitrate | |
[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
73.5°C | |
White | |
99.999% (Metals basis excluding Hg) | |
MFCD00003419 | |
Soluble in water,methanol,ethanol,and ethylene glycol. | |
[Al+3].[O-][N+]([O-])=O.[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
212.99 | |
212.99 (Anhydrous) | |
Hygroscopic | |
Aluminum nitrate hydrate |
25838-59-9 | |
Crystalline | |
AlN3O9 | |
aluminum nitrate hydrate, al.3no3.h2o, aluminum nitrate hydrate, puratronic, aluminum 3+ hydrate trinitrate | |
JLDSOYXADOWAKB-UHFFFAOYSA-N | |
aluminium(3+) trinitrate | |
22078336 | |
Odorless | |
White crystalline |
Safety and Handling
P210-P220-P221-P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362-P370+P378q-P501c
H272-H315-H319
missing translation for 'dotInformation' : DOT Class: 5.1, Packaging Group: III
missing translation for 'einecsNumber' : 236-751-8
missing translation for 'rtecsNumber' : BD1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
- Aluminum nitrate hydrate, Puratronic
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only