missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum nitrate nonahydrate, ACS, 98.0-102.0%, Thermo Scientific Chemicals
missing translation for 'orderingAttributeHoverText'
Quantity:
50 g
250 g
1 kg
5 x 1 kg
Descripción
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Especificaciones
Especificaciones
Melting Point | 73.5°C (135° (decomposition)) |
Color | White |
Physical Form | Crystalline |
Quantity | 50 g |
Assay Percent Range | 98 -102% |
Solubility Information | Very soluble in water and alcohol. Slightly soluble in acetone. Insoluble in ethyl acetate and pyridine. |
Formula Weight | 375.13 (212.99 Anhydrous) |
Grade | ACS |
Odor | Odorless |
Sensitivity | Hygroscopic |
Mostrar más |
Seguridad y manipulación
Nombre del producto
- Aluminum nitrate nonahydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido