Learn More
Aluminum nitrate nonahydrate, low mercury, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals
142.71 € - 1385.90 €
Chemical Identifiers
CAS | 7784-27-2 |
---|---|
Molecular Formula | AlH18N3O18 |
Molecular Weight (g/mol) | 375.129 |
MDL Number | MFCD00003419 |
InChI Key | SWCIQHXIXUMHKA-UHFFFAOYSA-N |
Synonym | aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic |
PubChem CID | 24567 |
IUPAC Name | aluminum;trinitrate;nonahydrate |
SMILES | [N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11364818
|
Thermo Scientific Alfa Aesar
043145.14 |
25 g |
142.71 €
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11374818
|
Thermo Scientific Alfa Aesar
043145.22 |
100 g |
375.87 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11384818
|
Thermo Scientific Alfa Aesar
043145.36 |
500 g |
1385.90 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum nitrate is a strong oxidizing agent. It is used in tanning leather, antiperspirants, corrosion inhibitors, extraction of uranium, petroleum refining, and as a nitrating agent. The nonahydrate and other hydrated aluminum nitrates have many applications. These salts are used to produce alumina for preparation of insulating papers, in cathode ray tube heating elements, and on transformer core laminates. The hydrated salts are also used for the extraction of actinide elements.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7784-27-2 | |
375.129 | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
24567 | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] |
AlH18N3O18 | |
MFCD00003419 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
aluminum;trinitrate;nonahydrate |
Specifications
73.5°C (135° (decomposition)) | |
Crystalline | |
MFCD00003419 | |
14,352 | |
Soluble in water (540mg/ml),ethanol,alcohol (very),and acetone (slightly). | |
[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].[N+](=O)([O-])[O-].O.O.O.O.O.O.O.O.O.[Al+3] | |
375.129 | |
375.13 (212.99 Anhydrous) | |
Puratronic™ | |
Hygroscopic | |
Aluminum nitrate nonahydrate, Low mercury |
7784-27-2 | |
AlH18N3O18 | |
UN1438 | |
aluminum nitrate nonahydrate, aluminum trinitrate nonahydrate, unii-8mc6621v1h, aluminum iii nitrate, nonahydrate 1:3:9, nitric acid aluminum salt, nitric acid, aluminum salt, nonahydrate 8ci,9ci, aluminum nitrate, acs, aluminum 3+ nonahydrate trinitronate, aluminum nitrate nonahydrate, low mercury, puratronic | |
SWCIQHXIXUMHKA-UHFFFAOYSA-N | |
aluminum;trinitrate;nonahydrate | |
24567 | |
99.999% | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H272-H315-H319
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
P280-P305+P351+P338-P310
H318
missing translation for 'dotInformation' : Transport Hazard Class: 5.1; Packing Group: III; Proper Shipping Name: ALUMINUM NITRATE
missing translation for 'einecsNumber' : 236-751-8
missing translation for 'rtecsNumber' : BD1050000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum nitrate nonahydrate
Signal Word
- Danger
Hazard Category
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
Hazard Statement
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P501-Dispose of contents/container to .
RUO – Research Use Only