Learn More
Aluminum nitride, N 32.0% min, Thermo Scientific Chemicals
44.62 € - 623.10 €
Chemical Identifiers
CAS | 24304-00-5 |
---|---|
Molecular Formula | AlN |
Molecular Weight (g/mol) | 40.99 |
MDL Number | MFCD00003429 |
InChI Key | PIGFYZPCRLYGLF-UHFFFAOYSA-N |
Synonym | aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c |
PubChem CID | 90455 |
ChEBI | CHEBI:50884 |
SMILES | N#[Al] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11367247
|
Thermo Scientific Alfa Aesar
011546.06 |
5 g |
44.62 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11377247
|
Thermo Scientific Alfa Aesar
011546.18 |
50 g |
62.81 €
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11357247
|
Thermo Scientific Alfa Aesar
011546.30 |
250 g |
217.08 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11387247
|
Thermo Scientific Alfa Aesar
011546.A1 |
1 kg |
623.10 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum nitride possesses very high thermal conductivity and effectively used in the ceramic industry. It finds application in deep ultraviolet optoelectronics, steel production, dielectric layers in optical storage media, chip carriers and as a crucible to grow crystals of gallium arsenide. It is also used as a radio frequency filter, which finds application in mobile phones. Further, it is used as a circuit carrier in semiconductors. In addition to this, it is used as a heat-sink in light-emitting diode lighting technology.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
24304-00-5 | |
40.99 | |
PIGFYZPCRLYGLF-UHFFFAOYSA-N | |
90455 | |
N#[Al] |
AlN | |
MFCD00003429 | |
aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c | |
CHEBI:50884 |
Specifications
>2200°C | |
White | |
AlN | |
14,353 | |
Soluble in mineral acids. | |
N#[Al] | |
40.99 | |
CHEBI:50884 | |
N 32.0% min. | |
Moisture sensitive | |
Aluminum nitride |
24304-00-5 | |
Powder | |
MFCD00003429 | |
aluminum nitride, unii-7k47d7p3m0, aluminum nitride paste, aluminum nitride 50g, aluminum nitride grade a, aluminum nitride grade b, aluminum nitride grade c | |
PIGFYZPCRLYGLF-UHFFFAOYSA-N | |
azanylidynealumane | |
90455 | |
40.99 | |
Ammonia-like | |
3.26 g/mL |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'einecsNumber' : 246-140-8
missing translation for 'rtecsNumber' : BD1055000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Aluminum nitride
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.