missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caframo™ High Torque Overhead Stirrer, 220 V
High Torque Overhead Stirrer, 220 Volt offers speed and torque capabilities for wide range of volumes and viscosities.
Brand: Caframo™ BDC3030220U1
This item is not returnable.
View return policy
Description
- Stirrers handle high-volume/high-viscosity stirring as easily as homogenizing and dispersing - especially useful for continuous use
- User-friendly keypad permits precise speed adjustment and digital LCD indicates rpm or torque to within ±1%
- Electronic controls permit speed adjustment to single-digit increments and maintain set speed within 1% as solution viscosity changes
- Totally Enclosed Metal Housing (TENV)
- Rugged, sealed metal housing encloses the high-performance brushless DC motor with automatic overload protection; and two-speed transmission that electronically shifts between high-torque and high-speed range - actually transferring motor's power to chuck with 95% efficiency
- Chuck is adjustable up to 10 mm (3/8 in.)
- Chuck guard protects chuck from splashes and prevents accidental movement of chuck
- Includes support rod, chuck, chuck key, chuck guard, UK and EU cord sets, and printed manual
- Stand, clamp and stirring impeller are sold separately
Specifications
20 to 3000 rpm | |
LED | |
9 in. | |
13.25 in. | |
230 V, 50/60 Hz | |
EU/UK/AUS/CN | |
Chemical Mixing in a Fume Hood or Enclosed Workstation, Pharma, Suspensions, Medical Labs, Serum Prep, Blood Product Mixing and Reactions | |
Aluminum | |
Brushless DC | |
Universal Stirrer |
60 L | |
220/240 V | |
22.9 cm | |
33.65 cm | |
50/60 Hz | |
CSA, CE, IEC, EMC RoHS, WEE | |
Clear chemical resistant hinged chuck guard | |
Universal | |
341 Ncm | |
50,000 mPas |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Caframo™ High Torque Overhead Stirrer, 220 V > High Torque Overhead Stirrer; Model: Universal; Speed: 20 to 600 (Low), 600 to 3000 (High); Capacity: 60 L; Voltage: 220 V
Spot an opportunity for improvement?Share a Content Correction