missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, 99% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
It finds applications in catalyst support, chemical sensors, as absorbent for gases and water vapors, manufacturing of abrasives, refractories, ceramics, electrical insulators and resistors, paper, crucibles, as chromatographic matrix, in fluxes, light bulbs, heat-resistant fiber. Heavy quantity of Aluminum oxide is used in the production of aluminum and a component of clays.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Boiling Point | 2980°C |
Melting Point | 2045°C |
Physical Form | Fine Powder |
Quantity | 500 g |
Assay Percent Range | 99% (metals basis) |
Solubility Information | Aluminum oxide is insoluble in water,but slightly soluble in acid and alkali. |
IUPAC Name | dialuminum;oxygen(2-) |
Formula Weight | 101.96 |
Refractive Index | 1.768 |
Density | 3.965 g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'