missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, acidic, HPLC Flash Grade, Thermo Scientific Chemicals

Product Code. 15452587
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Boiling Point 2980°C
Melting Point 2045°C
Color White
Physical Form Powder
Quantity 100 g
Linear Formula Al2O3
Merck Index 14,356
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade HPLC Flash
Odor Odorless
Refractive Index 1.765
Chemical Name or Material Aluminum oxide, Acidic
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.