missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, activated, acidic, Brockmann Grade I, 58 angstroms, Thermo Scientific Chemicals

Product Code. 11377177
Click to view available options
Quantity:
250 g
1 kg
5 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Acidic; S.A. 150m2/g

Aluminum oxide is used for the separation of both inorganic anions and acidic organic molecules such as acidic amino acids, aromatic acids and carboxylic acids. It is essential in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. Aluminum oxide is a source of aluminum in reactions, an abrasive agent, and as a refractory material. Also Aluminum oxide can act as a catalyst in a variety of reactions such as the claus process and in the dehydration of alcohols to alkenes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]

Specifications

Melting Point 2045°C
Color White
Physical Form Powder
Quantity 250 g
Solubility Information Insoluble in water.
IUPAC Name dialuminum;oxygen(2-)
Formula Weight 101.96
Grade Brockmann
Chemical Name or Material Aluminum oxide, Brockmann Grade I

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'