Learn More
Aluminum oxide, alpha-phase, 99.95% min (metals basis), Thermo Scientific Chemicals
39.40 € - 367.83 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11309188
|
Thermo Scientific Alfa Aesar
042573.22 |
100 g |
39.40 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11399178
|
Thermo Scientific Alfa Aesar
042573.36 |
500 g |
128.64 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11319188
|
Thermo Scientific Alfa Aesar
042573.A3 |
2 kg |
367.83 €
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aluminum oxide is widely used in a variety of plastics, rubber, ceramics, refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire, yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamp as far-infrared emission and thermal insulation materials. It is useful for the C single crystal detectors. It plays an important role for the hydrogenation of aromatic hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
2980°C | |
1344-28-1 | |
Powder | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
Odorless | |
Aluminum oxide |
2045°C | |
White | |
99.95% min. (Metals basis) | |
MFCD00003424 | |
Slowly soluble in aqueous alkali solutions-forming hydroxides. Insoluble in water and in non-polar organic solvents. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
1.765 |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only