missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum oxide, basic, HPLC Flash Grade, Thermo Scientific Chemicals
68.34 € - 231.15 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15422587
|
Thermo Scientific Alfa Aesar
045949.30 |
250 g |
68.34 €
250g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15432587
|
Thermo Scientific Alfa Aesar
045949.A1 |
1 kg |
231.15 €
1kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Specifications
2980°C | |
1344-28-1 | |
Powder | |
Al2O3 | |
14,356 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminum;oxygen(2-) | |
9989226 | |
HPLC Flash | |
1.765 |
2045°C | |
White | |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
Odorless | |
Aluminum oxide, Basic |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only