missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, gamma-phase, nanopowder, 99+%, Thermo Scientific Chemicals

59.60 € - 633.15 €

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11460001
View Documents
Thermo Scientific Alfa Aesar
044757.14
25 g
59.60 €
25g
Please to purchase this item. Need a web account? Register with us today!
11470001
View Documents
Thermo Scientific Alfa Aesar
044757.22
100 g
166.83 €
100g
Please to purchase this item. Need a web account? Register with us today!
11480001
View Documents
Thermo Scientific Alfa Aesar
044757.36
500 g
633.15 €
500g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum oxide is used in protein extraction as a component in the preparation of tissues. It is also used as a grinding or blending agent. It can act as a catalyst in a variety of reactions such as the Claus process. It is used in the production of aluminum, manufacture of abrasives, refractories, ceramics, electrical insulators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
Al2O3
MFCD00003424
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
[O--].[O--].[O--].[Al+3].[Al+3]

Specifications

1344-28-1
Powder
Al2O3
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminium(3+) trioxidandiide
9989226
Odorless
White
>99%
MFCD00003424
Insoluble in water.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Aluminum oxide
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only