Learn More
Aluminum oxide, NanoDur™ AL-0405, 99.5%, Thermo Scientific Chemicals
S. A. 32-40m2/g
62.30 € - 271.00 €
Chemical Identifiers
CAS | 1344-28-1 |
---|---|
Molecular Formula | Al2O3 |
Molecular Weight (g/mol) | 101.96 |
MDL Number | MFCD00003424 |
InChI Key | PNEYBMLMFCGWSK-UHFFFAOYSA-N |
Synonym | aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum |
PubChem CID | 9989226 |
IUPAC Name | dialuminium(3+) trioxidandiide |
SMILES | [O--].[O--].[O--].[Al+3].[Al+3] |
Description
NanoDur™ aluminum oxide is designed for coatings where transparency is of utmost importance. NanoDur™ aluminum oxide is especially suited for incorporation into coating systems, greatly enhancing abrasion resistance with minimal effects on cla..Alumium oxide is widely used in a variety of plastics, rubber, ceramics and refractory products for reinforcement toughening. It is also useful to produce artificial ruby, sapphire and yttrium aluminum garnet. It is useful in the field of fiber fabric products and high pressure sodium lamps as far-infrared emission and thermal insulation materials. It plays an important role for the hydrogenation of aromatic hydrocarbons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1344-28-1 | |
101.96 | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
9989226 | |
[O--].[O--].[O--].[Al+3].[Al+3] |
Al2O3 | |
MFCD00003424 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
dialuminium(3+) trioxidandiide |
Specifications
2980°C | |
1344-28-1 | |
Powder | |
Al2O3 | |
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum | |
PNEYBMLMFCGWSK-UHFFFAOYSA-N | |
dialuminium(3+) trioxidandiide | |
9989226 | |
Odorless | |
White powder | |
Aluminum oxide, Sold in collaboration with Nanophase Technologies Corporation |
2045°C | |
White | |
99.5% | |
MFCD00003424 | |
Slightly soluble in strong acid and alkaline solution. Insoluble in water. | |
[O--].[O--].[O--].[Al+3].[Al+3] | |
101.96 | |
101.96 | |
1.768 | |
3.965g/cm3 |
Safety and Handling
missing translation for 'einecsNumber' : 215-691-6
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only