missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide, neutral, HPLC Flash Grade, Thermo Scientific Chemicals

70.45 € - 218.09 €

Chemical Identifiers

CAS 1344-28-1
Molecular Formula Al2O3
Molecular Weight (g/mol) 101.96
MDL Number MFCD00003424
InChI Key PNEYBMLMFCGWSK-UHFFFAOYSA-N
Synonym aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PubChem CID 9989226
IUPAC Name dialuminum;oxygen(2-)
SMILES [O--].[O--].[O--].[Al+3].[Al+3]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15472587
View Documents
Thermo Scientific Alfa Aesar
045985.30
250 g
70.45 €
250g
Please to purchase this item. Need a web account? Register with us today!
15482587
View Documents
Thermo Scientific Alfa Aesar
045985.A1
1 kg
218.09 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Aluminum oxide is used as an adsorbent, desiccating agent, and catalyst, and in the manufacture of dental cements and refractories. Used in protein extraction as a component in the preparation of tissues. Used in bushings, spindles, lapping, polishing, refractory coating for investment casting shells, decorative non-skid tile, and pressure blasting. It is also used in varnishes, for textile impregnation, in cosmetics and as an intermediate in pharmaceutical production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1344-28-1
101.96
PNEYBMLMFCGWSK-UHFFFAOYSA-N
9989226
[O--].[O--].[O--].[Al+3].[Al+3]
Al2O3
MFCD00003424
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
dialuminum;oxygen(2-)

Specifications

2980°C
1344-28-1
Al2O3
MFCD00003424
Slightly soluble in alkaline solutions. Insoluble in water.
[O--].[O--].[O--].[Al+3].[Al+3]
101.96
101.96
Plastic bottle
Aluminum oxide
2045°C
Powder
Al2O3
aluminum oxide, alpha-alumina, fasertonerde, abramant, abramax, abrarex, abrasit, aloxite, alundum
PNEYBMLMFCGWSK-UHFFFAOYSA-N
dialuminum;oxygen(2-)
9989226
HPLC Flash
1.765
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.