missing translation for 'onlineSavingsMsg'
Learn More

Aluminum oxide substrate, 10x10x0.5mm, polished two sides, A plane, Thermo Scientific Chemicals

89.90 € - 342.00 €

Specifications

Chemical Name or Material Aluminum Oxide Substrate, A Plane
Name Note Polished Two Sides
Molecular Formula Al2O3
MDL Number MFCD00003424
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11394128
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041426.KT
1 Ea.
89.90 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11384128
View Documents Promotion Details
Thermo Scientific Alfa Aesar
041426.KU
5 Ea.
342.00 €
Pack of 5
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Aluminum Oxide Substrate, A Plane
Polished Two Sides
Al2O3
MFCD00003424
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only