missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, C Plane, Thermo Scientific™

188.00 € - 718.00 €

Specifications

Chemical Name or Material Aluminum Oxide Substrate, C Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424
Solubility Information Soluble in hexane and toluene. Insoluble in water and diethyl ether.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11319348
View Documents
Thermo Scientific Alfa Aesar
043248.KT
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11309348
View Documents
Thermo Scientific Alfa Aesar
043248.KU
5 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum Oxide Substrate, C Plane
Al2O3
Soluble in hexane and toluene. Insoluble in water and diethyl ether.
Polished One Side
MFCD00003424
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, C Plane, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.