missing translation for 'onlineSavingsMsg'
Learn More

Aluminum Oxide Substrate, 12.7 x 12.7 x 1mm, Polished One Side, M Plane, Thermo Scientific™

183.00 € - 696.00 €

Specifications

Chemical Name or Material Aluminum Oxide Substrate, M Plane
Name Note Polished One Side
Molecular Formula Al2O3
MDL Number MFCD00003424
Solubility Information Soluble in hexane and toluene. Insoluble in water and diethyl ether.
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11369338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043246.KT
1 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11379338
View Documents Promotion Details
Thermo Scientific Alfa Aesar
043246.KU
5 Ea. This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum Oxide Substrate, M Plane
Al2O3
Soluble in hexane and toluene. Insoluble in water and diethyl ether.
Polished One Side
MFCD00003424
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 215-691-6

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications