missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aluminum perchlorate nonahydrate, Reagent Grade, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
Description
Aluminum perchlorate nonahydrate is used as an intermediate in both pharmaceutical industry and chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 82°C |
Physical Form | Crystalline |
Quantity | 100 g |
UN Number | UN1481 |
Solubility Information | Soluble in water. |
Formula Weight | 487.46 (325.33 Anhydrous) |
Grade | Reagent |
Sensitivity | Hygroscopic |
Chemical Name or Material | Aluminum perchlorate nonahydrate |
Safety and Handling
Product Identifier
- Aluminum perchlorate nonahydrate
- Danger
- Oxidising solid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
- H272-May intensify fire; oxidiser.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P220-Keep/Store away from clothing/combustible materials.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction