Learn More
Aluminum phosphate, 97%, Thermo Scientific Chemicals
Description
As a flux for ceramics, preparation of dental cement, cement with calcium sulfate and sodium silicate, special glasses, as gel in pharmacy.Aluminum phosphate is used as catalysts, molecular sieves, flux for ceramics and as a gel in pharmacy. It is employed in the preparation of dental cement, emollients and fire-resistant coating. It plays an important role as an anti-fouling agent in textile industry.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | >1500°C |
Color | White |
Physical Form | Powder |
Quantity | 250 g |
Assay Percent Range | 97% |
Solubility Information | Insoluble in water and acetic acid. Slightly soluble in concentrated hydrochloric acid and nitric acid.Insoluble in water. |
Formula Weight | 121.95 |
Appearance | White powder |
Density | 2.566 g/cm3 |
Chemical Name or Material | Aluminum phosphate |
Safety and Handling
- Aluminum phosphate
- Danger
- Skin corrosion/irritation Category 1 B
- H314-Causes severe skin burns and eye damage.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.
- P501-Dispose of contents/container to .
RUO – Research Use Only