missing translation for 'onlineSavingsMsg'
Learn More

Aluminum phosphate, Puratronic™, 99.99% (metals basis), Thermo Scientific Chemicals

Product Code. 11300819
Click to view available options
Quantity:
5 g
25 g

Aluminum phosphate is used in the production of glass, mixture of ceramic, waterproofing concrete and dental cements, in cosmetics as a emollient, flame retardant, catalyst in organic synthesis, fireproofing textile, pharmaceuticals and dyes. It also used in many adhesives, binders and cements.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7784-30-7
Molecular Formula AlO4P
Molecular Weight (g/mol) 121.951
MDL Number MFCD00003430
InChI Key ILRRQNADMUWWFW-UHFFFAOYSA-K
Synonym aluminum phosphate, aluminum monophosphate, phosphalugel, phosphaljel, monoaluminum phosphate, aluminum acid phosphate, aluphos, aluminophosphoric acid, aluminiumphosphat german, aluminum phosphate 1:1
PubChem CID 64655
IUPAC Name aluminum;phosphate
SMILES [O-]P(=O)([O-])[O-].[Al+3]

Specifications

Melting Point >1500°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.99% (Metal basis)
Solubility Information Soluble in concentrated hydrochloric acid,alcohol and nitric acid. Insoluble in water.
Formula Weight 121.95
Appearance White powder
Density 2.566 g/cm3
Chemical Name or Material Aluminum phosphate
compliance-icons
Product Identifier
  • Aluminum phosphate, Puratronic
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'