missing translation for 'onlineSavingsMsg'
Learn More

Aluminum plate, alloy 6061, 2.3mm (0.09 in.) thick, Thermo Scientific™

29.00 € - 61.80 €

Specifications

Chemical Name or Material Aluminum plate, Alloy 6061
Name Note 2.3mm (0.09 in.) thick
MDL Number MFCD00134029
Solubility Information Insoluble in water.
Odor Odorless
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15422197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045193.GT
150 x 150 mm This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
15432197
View Documents Promotion Details
Thermo Scientific Alfa Aesar
045193.HB
300 x 300 mm
61.80 €
Each
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Aluminum plate, Alloy 6061
MFCD00134029
Odorless
2.3mm (0.09 in.) thick
Insoluble in water.
Plate
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications