missing translation for 'onlineSavingsMsg'
Learn More

Aluminum potassium sulfate dodecahydrate, Puriss., meets analytical spec. of BP, Ph. Eur., 99.0-100.5% (calc. on dry substance), Honeywell Fluka™
BIOPHARMA

Catalog Number 15651160
97.74 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.


Click to view available options
Quantity:
1 kg
6 x 1 kg
This item is not returnable. View return policy
7784-24-9
AlH24KO20S2
474.372
MFCD00149143
GNHOJBNSNUXZQA-UHFFFAOYSA-J
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
62667
CHEBI:86465
aluminum;potassium;disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]
This item is not returnable. View return policy
7784-24-9
AlH24KO20S2
474.372
MFCD00149143
GNHOJBNSNUXZQA-UHFFFAOYSA-J
kalinite, aluminum potassium sulfate dodecahydrate, potassium alum dodecahydrate, potash alum dodecahydrate, aluminum potassium disulfate dodecahydrate, unii-1l24v9r23s, alum, potassium, dodecahydrate, potassium aluminum sulfate dodecahydrate, aluminum potassium sulfate dodecahydrate, potassium aluminum disulfate dodecahydrate
62667
CHEBI:86465
aluminum;potassium;disulfate;dodecahydrate
O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Al+3].[K+]

Puriss., meets analytical specification of BP, European Pharmacopoeia, 99.0-100.5% (calc. on dry substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY
Quantity 1 kg
Linear Formula AlK(SO4)2 · 12H2O
UN Number NONH for all modes of transport
Formula Weight 474.39g/mol
Percent Purity 99 to 100.5% (calc. to the dried substance)
Grade puriss.
Packaging Poly bottle
Density 1.757g/mL at 25°C
Chemical Name or Material Aluminum Potassium Sulfate Dodecahydrate, Meets analytical specification of Ph.Eur.BP, NF, FCC
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'